Tisk…

2-Fluorobenzenesulphonyl Fluoride

Dodavatel: Apollo Scientific

PC510011-25G PC510011-5G
APOSPC510011-25GEA 26090 CZK
APOSPC510011-25G APOSPC510011-5G
2-Fluorobenzenesulphonyl Fluoride
2-Fluorobenzenesulphonyl Fluoride
Vzorec: C₆H₄F₂O₂S
MW: 178,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18375235
CAS číslo: 52200-99-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR