Tisk…

3-Fluoro-4-pyrrolidinobenzoic acid

Dodavatel: Apollo Scientific

PC510009-1G PC510009-5G
APOSPC510009-1GEA 4670 CZK
APOSPC510009-1G APOSPC510009-5G
3-Fluoro-4-pyrrolidinobenzoic acid
3-Fluoro-4-pyrrolidinobenzoic acid
Vzorec: C₁₁H₁₂FNO₂
Storage Temperature: Ambient
MDL Number: MFCD11156505
CAS číslo: 1021243-16-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR