4-(4-Ethyl-1-piperazinyl)-2-fluoroaniline hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
PC510007-1G
PC510007-5G
APOSPC510007-1GEA
7280
CZK
APOSPC510007-1G
APOSPC510007-5G
4-(4-Ethyl-1-piperazinyl)-2-fluoroaniline hydrochloride
4-(4-Ethyl-1-piperazinyl)-2-fluoroaniline hydrochloride
Vzorec:
C₁₂H₁₈FN₃·ClH Storage Temperature: Ambient |
MDL Number:
MFCD13191588 CAS číslo: 1197193-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...