5-(4-Fluorobenzyl)thiazolidine-2,4-dione
Dodavatel: Apollo Scientific
PC510004-5G
PC510004-1G
APOSPC510004-5GEA
41330
CZK
APOSPC510004-5G
APOSPC510004-1G
5-(4-Fluorobenzyl)thiazolidine-2,4-dione
5-(4-Fluorobenzyl)thiazolidine-2,4-dione
Vzorec:
C₁₀H₈FNO₂S Storage Temperature: Ambient |
MDL Number:
MFCD12024822 CAS číslo: 291536-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...