4-Fluoro-5-morpholino-2-nitrobenzoic acid
Dodavatel: Apollo Scientific
PC510003-1G
PC510003-5G
APOSPC510003-1GEA
7280
CZK
APOSPC510003-1G
APOSPC510003-5G
4-Fluoro-5-morpholino-2-nitrobenzoic acid
4-Fluoro-5-morpholino-2-nitrobenzoic acid
Vzorec:
C₁₁H₁₁FN₂O₅ Storage Temperature: Ambient |
MDL Number:
MFCD11656825 CAS číslo: 1033778-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...