Tisk…

Methyl 2-Amino-5-fluoro-4-(1-pyrazolyl)benzoate

Dodavatel: Apollo Scientific

PC510001-1G PC510001-5G
APOSPC510001-1GEA 5530 CZK
APOSPC510001-1G APOSPC510001-5G
Methyl 2-Amino-5-fluoro-4-(1-pyrazolyl)benzoate
Methyl 2-Amino-5-fluoro-4-(1-pyrazolyl)benzoate
Vzorec: C₁₁H₁₀FN₃O₂
MW: 235,214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656805
CAS číslo: 1186663-61-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR