Tisk…
2,4-Dimethoxy-3-fluorobenzoic acid 95%
Dodavatel: Apollo Scientific
Synonyms: 3-Fluoro-2,4-dimethoxybenzoic acid
PC50953-1G PC50953-250MG
APOSPC50953-1GEA 16340 CZK
APOSPC50953-1G APOSPC50953-250MG
2,4-Dimethoxy-3-fluorobenzoic acid 95%
2,4-Dimethoxy-3-fluorobenzoic acid
Vzorec: C₉H₉FO₄
MW: 200,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24498657
CAS číslo: 1018451-10-9