Tisk…

1-Bromo-5-chloro-3,4-difluoro-2-iodobenzene 95%

Dodavatel: Apollo Scientific

PC50949-1G PC50949-250MG PC50949-5G
APOSPC50949-1GEA 14880 CZK
APOSPC50949-1G APOSPC50949-250MG APOSPC50949-5G
1-Bromo-5-chloro-3,4-difluoro-2-iodobenzene 95%
1-Bromo-5-chloro-3,4-difluoro-2-iodobenzene
Vzorec: C₆HBrClF₂I
MW: 353,33 g/mol
Storage Temperature: Ambient
CAS číslo: 2451256-38-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR