5-Bromo-4-chloro-8-(trifluoromethyl)quinoline
Dodavatel: Apollo Scientific
|
Varování
|
PC508136-1G
PC508136-250MG
APOSPC508136-1GEA
8010
CZK
APOSPC508136-1G
APOSPC508136-250MG
5-Bromo-4-chloro-8-(trifluoromethyl)quinoline
5-Bromo-4-chloro-8-(trifluoromethyl)quinoline
Vzorec:
C₁₀H₄BrClF₃N MW: 310,498 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28010389 CAS číslo: 1065093-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...