Tisk…

8-Bromo-4-chloro-5-(trifluoromethyl)quinoline

Dodavatel: Apollo Scientific
Varování

PC508133-1G PC508133-250MG
APOSPC508133-1GEA 8010 CZK
APOSPC508133-1G APOSPC508133-250MG
8-Bromo-4-chloro-5-(trifluoromethyl)quinoline
8-Bromo-4-chloro-5-(trifluoromethyl)quinoline
Vzorec: C₁₀H₄BrClF₃N
MW: 310,498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28010390
CAS číslo: 1065093-18-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR