Tisk…

6-Bromo-4-chloro-8-(trifluoromethyl)quinoline

Dodavatel: Apollo Scientific
Varování

PC508130-1G PC508130-250MG
APOSPC508130-1GEA 14880 CZK
APOSPC508130-1G APOSPC508130-250MG
6-Bromo-4-chloro-8-(trifluoromethyl)quinoline
6-Bromo-4-chloro-8-(trifluoromethyl)quinoline
Vzorec: C₁₀H₄BrClF₃N
Storage Temperature: Ambient
MDL Number: MFCD28010396
CAS číslo: 1065093-30-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR