8-Bromo-4-chloro-6-fluoroquinoline
Dodavatel: Apollo Scientific
|
Varování
|
PC508125-1G
PC508125-250MG
APOSPC508125-1GEA
14060
CZK
APOSPC508125-1G
APOSPC508125-250MG
8-Bromo-4-chloro-6-fluoroquinoline
8-Bromo-4-chloro-6-fluoroquinoline
Vzorec:
C₉H₄BrClFN MW: 260,49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09861347 CAS číslo: 1019016-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...