Tisk…

8-Chloro-7-fluoroquinolin-4(1H)-one

Dodavatel: Apollo Scientific
Varování

PC508123-1G PC508123-250MG
APOSPC508123-1GEA 6850 CZK
APOSPC508123-1G APOSPC508123-250MG
8-Chloro-7-fluoroquinolin-4(1H)-one
8-Chloro-7-fluoroquinolin-4(1H)-one
Vzorec: C₉H₅ClFNO
MW: 197,594 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31745743
CAS číslo: 1065092-33-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR