Tisk…

8-Bromo-7-(trifluoromethyl)quinolin-4(1H)-one

Dodavatel: Apollo Scientific
Varování

PC508122-1G PC508122-250MG
APOSPC508122-1GEA 12200 CZK
APOSPC508122-1G APOSPC508122-250MG
8-Bromo-7-(trifluoromethyl)quinolin-4(1H)-one
8-Bromo-7-(trifluoromethyl)quinolin-4(1H)-one
Vzorec: C₁₀H₅BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD31745742
CAS číslo: 2366994-71-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR