Tisk…

6-Chloro-8-(trifluoromethyl)quinolin-4(1H)-one

Dodavatel: Apollo Scientific
Varování

PC508113-1G PC508113-250MG
APOSPC508113-1GEA 6850 CZK
APOSPC508113-1G APOSPC508113-250MG
6-Chloro-8-(trifluoromethyl)quinolin-4(1H)-one
6-Chloro-8-(trifluoromethyl)quinolin-4(1H)-one
Vzorec: C₁₀H₅ClF₃NO
MW: 247,601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21607792
CAS číslo: 1065092-54-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR