8-Bromo-6-(trifluoromethyl)quinolin-4(1H)-one
Dodavatel: Apollo Scientific
|
Varování
|
PC508106-1G
PC508106-250MG
APOSPC508106-1GEA
6850
CZK
APOSPC508106-1G
APOSPC508106-250MG
8-Bromo-6-(trifluoromethyl)quinolin-4(1H)-one
8-Bromo-6-(trifluoromethyl)quinolin-4(1H)-one
Vzorec:
C₁₀H₅BrF₃NO Storage Temperature: Ambient |
MDL Number:
MFCD31745731 CAS číslo: 1065092-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...