6-Bromo-2,3,4-trifluorobenzonitrile 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC50544-1G
PC50544-250MG
PC50544-5G
APOSPC50544-1GEA
1660
CZK
APOSPC50544-1G
APOSPC50544-250MG
APOSPC50544-5G
6-Bromo-2,3,4-trifluorobenzonitrile 95%
6-Bromo-2,3,4-trifluorobenzonitrile
Vzorec:
C₇HBrF₃N MW: 235,989 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32264074 CAS číslo: 2407339-61-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...