Tisk…

1-[3-(Trifluoromethyl)phenyl]imidazolidin-2-one

Dodavatel: Apollo Scientific
Varování

PC50543-1G PC50543-250MG
APOSPC50543-1GEA 13230 CZK
APOSPC50543-1G APOSPC50543-250MG
1-[3-(Trifluoromethyl)phenyl]imidazolidin-2-one
1-[3-(Trifluoromethyl)phenyl]imidazolidin-2-one
Vzorec: C₁₀H₉F₃N₂O
MW: 230,186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03787197
CAS číslo: 14088-95-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR