Tisk…

3-Bromo-1,2,4-trifluoro-5-iodobenzene 95%

Dodavatel: Apollo Scientific

PC50479-1G PC50479-250MG
APOSPC50479-1GEA 14880 CZK
APOSPC50479-1G APOSPC50479-250MG
3-Bromo-1,2,4-trifluoro-5-iodobenzene 95%
3-Bromo-1,2,4-trifluoro-5-iodobenzene
Vzorec: C₆HBrF₃I
MW: 336,876 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32662414
CAS číslo: 2451256-37-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR