Tisk…

3-Bromo-2,4,5-trifluoroaniline ≥98%

Dodavatel: Apollo Scientific

PC50478-100MG PC50478-1G PC50478-250MG PC50478-5G
APOSPC50478-100MGEA 417 CZK
APOSPC50478-100MG APOSPC50478-1G APOSPC50478-250MG APOSPC50478-5G
3-Bromo-2,4,5-trifluoroaniline ≥98%
3-Bromo-2,4,5-trifluoroaniline
Vzorec: C₆H₃BrF₃N
MW: 225,994 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30530500
CAS číslo: 2055841-42-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR