Tisk…

2,4-Dichloro-6-fluorophenol 97%

Dodavatel: Apollo Scientific

PC50442-1G PC50442-5G
APOSPC50442-1GEA 7110 CZK
APOSPC50442-1G APOSPC50442-5G
2,4-Dichloro-6-fluorophenol 97%
2,4-Dichloro-6-fluorophenol
Vzorec: C₆H₃Cl₂FO
MW: 180,992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405440
CAS číslo: 344-21-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR