Tisk…

4-Ethoxy-3,5-difluorobromobenzene 98%

Dodavatel: Apollo Scientific

Synonyms: 5-Bromo-2-ethoxy-1,3-difluorobenzene

PC50441-10G PC50441-1G PC50441-5G
APOSPC50441-10GEA 16340 CZK
APOSPC50441-10G APOSPC50441-1G APOSPC50441-5G
4-Ethoxy-3,5-difluorobromobenzene 98%
4-Ethoxy-3,5-difluorobromobenzene
Vzorec: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917873
CAS číslo: 115467-04-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR