Tisk…

3-Chloro-5-(trifluoromethoxy)bromobenzene 97%

Dodavatel: Apollo Scientific

Synonyms: 1-Bromo-3-chloro-5-(trifluoromethoxy)benzene

PC50439-10G PC50439-1G PC50439-5G PC50439-25G
APOSPC50439-10GEA 16370 CZK
APOSPC50439-10G APOSPC50439-1G APOSPC50439-5G APOSPC50439-25G
3-Chloro-5-(trifluoromethoxy)bromobenzene 97%
3-Chloro-5-(trifluoromethoxy)bromobenzene
Vzorec: C₇H₃BrClF₃O
MW: 275,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856023
CAS číslo: 1417567-41-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR