4-Fluoro-3-methyl-2-(trifluoromethyl)bromobenzene 98%
Dodavatel: Apollo Scientific
PC50432-10G
PC50432-1G
PC50432-5G
APOSPC50432-10GEA
16340
CZK
APOSPC50432-10G
APOSPC50432-1G
APOSPC50432-5G
4-Fluoro-3-methyl-2-(trifluoromethyl)bromobenzene 98%
4-Fluoro-3-methyl-2-(trifluoromethyl)bromobenzene
Vzorec:
C₈H₅BrF₄ MW: 257,023 g/mol Storage Temperature: Ambient |
CAS číslo:
2167351-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...