Tisk…

6-Chloro-2,3-difluorotoluene 98%

Dodavatel: Apollo Scientific

Synonyms: 2,3-Difluoro-6-chlorotoluene

PC50422-1G PC50422-250MG PC50422-5G
APOSPC50422-1GEA 8270 CZK
APOSPC50422-1G APOSPC50422-250MG APOSPC50422-5G
6-Chloro-2,3-difluorotoluene 98%
6-Chloro-2,3-difluorotoluene
Vzorec: C₇H₅ClF₂
MW: 162,564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922582
CAS číslo: 1208077-23-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR