4-Fluoro-3-methyl-5-(trifluoromethyl)nitrobenzene 97%
Dodavatel: Apollo Scientific
PC50421-10G
PC50421-1G
PC50421-5G
APOSPC50421-10GEA
16770
CZK
APOSPC50421-10G
APOSPC50421-1G
APOSPC50421-5G
4-Fluoro-3-methyl-5-(trifluoromethyl)nitrobenzene 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)nitrobenzene
Vzorec:
C₈H₅F₄NO₂ MW: 223,124 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28797683 CAS číslo: 1803832-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...