Tisk…

3-Chloro-5-(pentafluorosulphur)bromobenzene 98%

Dodavatel: Apollo Scientific

PC50419-10G PC50419-1G PC50419-5G
APOSPC50419-10GEA 53560 CZK
APOSPC50419-10G APOSPC50419-1G APOSPC50419-5G
3-Chloro-5-(pentafluorosulphur)bromobenzene 98%
3-Chloro-5-(pentafluorosulphur)bromobenzene
Vzorec: C₆H₃BrClF₅S
MW: 317,502 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32708472
CAS číslo: 2514941-99-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR