4-Bromo-2,5-difluorobenzamide
Dodavatel: Apollo Scientific
|
Varování
|
PC50405-1G
PC50405-250MG
APOSPC50405-1GEA
2240
CZK
APOSPC50405-1G
APOSPC50405-250MG
4-Bromo-2,5-difluorobenzamide
4-Bromo-2,5-difluorobenzamide
Vzorec:
C₇H₄BrF₂NO MW: 236.014 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847048 CAS číslo: 764648-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...