1-(3-Bromoprop-1-ynyl)-4-fluoro-benzene 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC50403-1G
PC50403-250MG
PC50403-5G
APOSPC50403-1GEA
14880
CZK
APOSPC50403-1G
APOSPC50403-250MG
APOSPC50403-5G
1-(3-Bromoprop-1-ynyl)-4-fluoro-benzene 98%
1-(3-Bromoprop-1-ynyl)-4-fluoro-benzene
Vzorec:
C₉H₆BrF Storage Temperature: Freezer |
MDL Number:
MFCD22482468 CAS číslo: 40274-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...