Tisk…

4-(Difluoromethyl)-2-(trifluoromethyl)anisole 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC50391-1G PC50391-250MG
APOSPC50391-1GEA 2570 CZK
APOSPC50391-1G APOSPC50391-250MG
4-(Difluoromethyl)-2-(trifluoromethyl)anisole 95%
4-(Difluoromethyl)-2-(trifluoromethyl)anisole
Vzorec: C₉H₇F₅O
Storage Temperature: Ambient
MDL Number: MFCD14698614
CAS číslo: 1214348-62-5
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR