Tisk…

7-Fluoro-2-oxo-2H-chromene-3-carboxylic acid

Dodavatel: Apollo Scientific
Varování

PC50388-1G PC50388-250MG
APOSPC50388-1GEA 9920 CZK
APOSPC50388-1G APOSPC50388-250MG
7-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
7-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
Vzorec: C₁₀H₅FO₄
MW: 208,143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26593782
CAS číslo: 1353870-09-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR