Tisk…

2-Bromo-3,5-difluorobenzyl alcohol 95%

Dodavatel: Apollo Scientific
Varování

PC50383-1G PC50383-250MG PC50383-5G
APOSPC50383-1GEA 9720 CZK
APOSPC50383-1G APOSPC50383-250MG APOSPC50383-5G
2-Bromo-3,5-difluorobenzyl alcohol 95%
2-Bromo-3,5-difluorobenzyl alcohol
Vzorec: C₇H₅BrF₂O
MW: 223.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28737913
CAS číslo: 1256081-83-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR