6-Fluorobenzofuran-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC50382-1G
PC50382-250MG
APOSPC50382-1GEA
10350
CZK
APOSPC50382-1G
APOSPC50382-250MG
6-Fluorobenzofuran-2-carboxylic acid
6-Fluorobenzofuran-2-carboxylic acid
Vzorec:
C₉H₅FO₃ MW: 180,133 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14708187 CAS číslo: 26018-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...