Tisk…

2-(3-Bromophenyl)-2,2-difluoroacetic acid

Dodavatel: Apollo Scientific
Nebezpečí

PC50325-1G PC50325-250MG PC50325-5G
APOSPC50325-1GEA 3910 CZK
APOSPC50325-1G APOSPC50325-250MG APOSPC50325-5G
2-(3-Bromophenyl)-2,2-difluoroacetic acid
2-(3-Bromophenyl)-2,2-difluoroacetic acid
Vzorec: C₈H₅BrF₂O₂
MW: 251,025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11167799
CAS číslo: 885068-76-8
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR