Tisk…

4-Bromo-2-(trifluoromethoxy)phenylisocyanate

Dodavatel: Apollo Scientific
Nebezpečí

PC503013-1G PC503013-25G PC503013-5G
APOSPC503013-1GEA 622 CZK
APOSPC503013-1G APOSPC503013-25G APOSPC503013-5G
4-Bromo-2-(trifluoromethoxy)phenylisocyanate
4-Bromo-2-(trifluoromethoxy)phenylisocyanate

May form small amount of sediment

Vzorec: C₈H₃BrF₃NO₂
MW: 282.014 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01320693
CAS číslo: 229957-06-6
UN: 2206
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR