3-Bromo-4-fluorophenylisocyanate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC503010-1G
PC503010-25G
PC503010-5G
PC503010-100G
APOSPC503010-1GEA
2480
CZK
APOSPC503010-1G
APOSPC503010-25G
APOSPC503010-5G
APOSPC503010-100G
3-Bromo-4-fluorophenylisocyanate
3-Bromo-4-fluorophenylisocyanate
May form small amount of sediment
Vzorec:
C₇H₃BrFNO MW: 216.007 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22557618 CAS číslo: 900574-93-8 UN: 2206 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...