Tisk…

2-Bromo-4-(trifluoromethoxy)phenylisocyanate

Dodavatel: Apollo Scientific
Nebezpečí

PC502997-1G PC502997-25G PC502997-5G
APOSPC502997-1GEA 622 CZK
APOSPC502997-1G APOSPC502997-25G APOSPC502997-5G
2-Bromo-4-(trifluoromethoxy)phenylisocyanate
2-Bromo-4-(trifluoromethoxy)phenylisocyanate

May form small amount of sediment

Vzorec: C₈H₃BrF₃NO₂
MW: 282.014 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32200433
CAS číslo: 848771-80-2
UN: 2206
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR