2-Bromo-3-fluorobenzyl fluoride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC502903-1G
PC502903-250MG
APOSPC502903-1GEA
17360
CZK
APOSPC502903-1G
APOSPC502903-250MG
2-Bromo-3-fluorobenzyl fluoride 95%
2-Bromo-3-fluorobenzyl fluoride
Vzorec:
C₇H₅BrF₂ Storage Temperature: Ambient |
MDL Number:
MFCD31634837 CAS číslo: 2244088-41-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...