2-Bromo-5-(trifluoromethyl)benzyl fluoride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC502902-1G
PC502902-250MG
APOSPC502902-1GEA
17360
CZK
APOSPC502902-1G
APOSPC502902-250MG
2-Bromo-5-(trifluoromethyl)benzyl fluoride 95%
2-Bromo-5-(trifluoromethyl)benzyl fluoride
Vzorec:
C₈H₅BrF₄ MW: 257.023 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31634836 CAS číslo: 2244088-55-7 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...