Tisk…

3-Bromo-4-(trifluoromethyl)benzyl fluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC502899-1G PC502899-250MG
APOSPC502899-1GEA 17360 CZK
APOSPC502899-1G APOSPC502899-250MG
3-Bromo-4-(trifluoromethyl)benzyl fluoride 95%
3-Bromo-4-(trifluoromethyl)benzyl fluoride
Vzorec: C₈H₅BrF₄
MW: 257,023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31634835
CAS číslo: 2244087-06-5
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR