Tisk…

tert-Butyl (R)-(1-phenyl-2-((trifluoromethyl)thio)ethyl)carbamate 95%

Dodavatel: Apollo Scientific
Varování

PC502897-1G PC502897-250MG
APOSPC502897-1GEA 1990 CZK
APOSPC502897-1G APOSPC502897-250MG
tert-Butyl (R)-(1-phenyl-2-((trifluoromethyl)thio)ethyl)carbamate 95%
tert-Butyl (R)-(1-phenyl-2-((trifluoromethyl)thio)ethyl)carbamate
Vzorec: C₁₄H₁₈F₃NO₂S
Storage Temperature: Ambient
MDL Number: MFCD31653061
CAS číslo: 2092899-90-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR