Tisk…

tert-Butyl (R)-(3-methyl-1-((trifluoromethyl)thio)butan-2-yl)carbamate 95%

Dodavatel: Apollo Scientific
Varování

PC502895-1G PC502895-250MG
APOSPC502895-1GEA 2650 CZK
APOSPC502895-1G APOSPC502895-250MG
tert-Butyl (R)-(3-methyl-1-((trifluoromethyl)thio)butan-2-yl)carbamate 95%
tert-Butyl (R)-(3-methyl-1-((trifluoromethyl)thio)butan-2-yl)carbamate
Vzorec: C₁₁H₂₀F₃NO₂S
MW: 287,342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31653058
CAS číslo: 2244064-17-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR