tert-Butyl (S)-(3-methyl-1-((trifluoromethyl)thio)butan-2-yl)carbamate 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC502894-1G
PC502894-250MG
APOSPC502894-1GEA
2650
CZK
APOSPC502894-1G
APOSPC502894-250MG
tert-Butyl (S)-(3-methyl-1-((trifluoromethyl)thio)butan-2-yl)carbamate 95%
tert-Butyl (S)-(3-methyl-1-((trifluoromethyl)thio)butan-2-yl)carbamate
Vzorec:
C₁₁H₂₀F₃NO₂S MW: 287,342 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31653057 CAS číslo: 2092899-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...