Tisk…

2-Bromo-3-nitrobenzyl fluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC502886-250MG
APOSPC502886-250MGEA 2280 CZK
APOSPC502886-250MG
2-Bromo-3-nitrobenzyl fluoride 95%
2-Bromo-3-nitrobenzyl fluoride
Vzorec: C₇H₅BrFNO₂
MW: 234,023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31634831
CAS číslo: 2244083-36-9
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR