Tisk…

3-Bromo-4-(trifluoromethoxy)benzyl bromide

Dodavatel: Apollo Scientific

PC502696-1G PC502696-5G
APOSPC502696-1GEA 7940 CZK
APOSPC502696-1G APOSPC502696-5G
3-Bromo-4-(trifluoromethoxy)benzyl bromide
3-Bromo-4-(trifluoromethoxy)benzyl bromide
Vzorec: C₈H₅Br₂F₃O
MW: 333.928 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561242
CAS číslo: 1011531-45-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR