Tisk…

5-Bromo-2-(trifluoromethoxy)benzyl alcohol 98%

Dodavatel: Apollo Scientific

PC502693-1G PC502693-5G
APOSPC502693-1GEA 3540 CZK
APOSPC502693-1G APOSPC502693-5G
5-Bromo-2-(trifluoromethoxy)benzyl alcohol 98%
5-Bromo-2-(trifluoromethoxy)benzyl alcohol
Vzorec: C₈H₆BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD03701031
CAS číslo: 685126-86-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR