Methyl 3-bromo-5-(trifluoromethoxy)benzoate
Dodavatel: Apollo Scientific
PC502691-1G
PC502691-5G
APOSPC502691-1GEA
1750
CZK
APOSPC502691-1G
APOSPC502691-5G
Methyl 3-bromo-5-(trifluoromethoxy)benzoate
Methyl 3-bromo-5-(trifluoromethoxy)benzoate
Vzorec:
C₉H₆BrF₃O₃ MW: 299.041 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09999511 CAS číslo: 1306763-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...