2-Bromo-6-(trifluoromethoxy)benzyl alcohol 95%
Dodavatel: Apollo Scientific
PC502688-1G
PC502688-5G
PC502688-10G
APOSPC502688-1GEA
3740
CZK
APOSPC502688-1G
APOSPC502688-5G
APOSPC502688-10G
2-Bromo-6-(trifluoromethoxy)benzyl alcohol 95%
2-Bromo-6-(trifluoromethoxy)benzyl alcohol
Vzorec:
C₈H₆BrF₃O₂ MW: 271,031 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22375105 CAS číslo: 1253189-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...