Methyl 3-chloro-2-fluorobenzoate 98%
Dodavatel: Apollo Scientific
PC502673-1G
PC502673-5G
APOSPC502673-1GEA
1410
CZK
APOSPC502673-1G
APOSPC502673-5G
Methyl 3-chloro-2-fluorobenzoate 98%
Methyl 3-chloro-2-fluorobenzoate
Vzorec:
C₈H₆ClFO₂ MW: 188,583 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06203787 CAS číslo: 773874-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...