Methyl 2-chloro-5-(trifluoromethyl)benzoate 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC502609-1G
PC502609-5G
PC502609-250MG
APOSPC502609-1GEA
1990
CZK
APOSPC502609-1G
APOSPC502609-5G
APOSPC502609-250MG
Methyl 2-chloro-5-(trifluoromethyl)benzoate 98%
Methyl 2-chloro-5-(trifluoromethyl)benzoate
Vzorec:
C₉H₆ClF₃O₂ MW: 238,591 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06203726 CAS číslo: 26107-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...